519642
2-Benzyloxybromobenzene
96%
Synonym(s):
Benzyl 2-bromophenyl ether
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
96%
form
solid
refractive index
n20/D >1.6060 (lit.)
functional group
bromo
phenyl
SMILES string
Brc1ccccc1OCc2ccccc2
InChI
1S/C13H11BrO/c14-12-8-4-5-9-13(12)15-10-11-6-2-1-3-7-11/h1-9H,10H2
InChI key
NBHAHMHUMMWFPJ-UHFFFAOYSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Utilization of a copper-catalyzed diaryl ether synthesis for the preparation of verbenachalcone.
Tetrahedron, 58(39), 7903-7910 (2002)
Self-assembly of tetraphenol and its complexes with aromatic diamines: Novel interpenetrating and noninterpenetrating organic assemblies.
Crystal Growth & Design, 5(4), 1575-1583 (2005)
Formation of interesting organic supramolecular structures in the solid-state self-assembly of triphenol adducts.
Crystal Growth & Design, 6(1), 150-160 (2006)
Synthesis of isotopically labelled salicylates.
Journal of Labelled Compounds & Radiopharmaceuticals, 18(4), 593-602 (1981)
Synthesis and solid-state self-assembly of polyphenols.
Crystal Growth & Design, 4(3), 553-561 (2004)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service