Skip to Content
Merck
All Photos(1)

Key Documents

519618

Sigma-Aldrich

2,2-Diethoxypropane

97%

Synonym(s):

Acetone diethyl acetal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C(OC2H5)2
CAS Number:
Molecular Weight:
132.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.389 (lit.)

bp

116-117 °C (lit.)

density

0.82 g/mL at 25 °C (lit.)

functional group

ether
ketal

storage temp.

2-8°C

SMILES string

CCOC(C)(C)OCC

InChI

1S/C7H16O2/c1-5-8-7(3,4)9-6-2/h5-6H2,1-4H3

InChI key

FGQLGYBGTRHODR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

46.4 °F - closed cup

Flash Point(C)

8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid chemical dehydration of plant material for light and electron microscopy with 2, 2-dimethoxypropane and 2, 2-diethoxypropane.
Lin CH, et al.
American Journal of Botany, 602-605 (1977)
Hydrolysis of D-(--)-Ethyl α-Benzoyloxypropionate and L-(--)-Ethyl α-Acetoxy-β-phenylpropionate by α-Chymotrypsin.
Cohen SG and Weinstein SY.
Journal of the American Chemical Society, 86(23), 5326-5330 (1964)
The Preparation of Some Ketals of Alkylacetylenes with the Higher Alcohols1.
Killian DB, et al.
Journal of the American Chemical Society, 58(1), 80-81 (1936)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service