Skip to Content
Merck
All Photos(1)

Key Documents

518646

Sigma-Aldrich

2-Amino-4-chloro-6-methoxypyrimidine

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6ClN3O
CAS Number:
Molecular Weight:
159.57
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

mp

168-171 °C (lit.)

functional group

chloro

SMILES string

COc1cc(Cl)nc(N)n1

InChI

1S/C5H6ClN3O/c1-10-4-2-3(6)8-5(7)9-4/h2H,1H3,(H2,7,8,9)

InChI key

VFEYBTFCBZMBAU-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 2-amino-4-chloro-6-methoxypyrimidine.
Zhu L, et al.
Journal of Zhejiang University. Science, 32(2), 87-88 (2000)
Christer B Aakeröy et al.
Inorganic chemistry, 44(14), 4983-4991 (2005-07-07)
The directed assembly of six different M(II) complexes (M = Cu, Co, and Ni) into infinite chains has been achieved by combining anionic chelating ligands (for controlling the coordination geometry) with bifunctional ligands containing a metal-coordinating pyridyl moiety and a
C W Klampfl et al.
Journal of chromatography. A, 911(2), 277-283 (2001-04-11)
This work describes the use of mixed-mode stationary phases which exhibit both strong ion-exchange (either cation-exchange, SCX, or anion-exchange, SAX) and reversed-phase chromatographic characteristics in capillary electrochromatographic separations of pyrimidine derivatives. Different packing materials, namely C6, SCX/C6 and SAX/C6, were
Z Cinar et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 116, 451-459 (2013-08-27)
The 2-amino-4-chloro-6-methoxypyrimidine abbreviated as ACMP have been investigated by both the experimental and theoretical methods; through this work we provide the essential fact about the structural and vibrational insights. The optimized molecular structure, atomic charges, vibrational frequencies and ultraviolet spectral

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service