Skip to Content
Merck
All Photos(1)

Key Documents

516031

Sigma-Aldrich

Methyl (S)-(−)-N-Z-aziridine-2-carboxylate

96%

Synonym(s):

(2S)-1,2-Aziridinedicarboxylic acid 2-methyl 1-(phenylmethyl) ester, (S)-N-Cbz-aziridine-2-carboxylic acid methyl ester, 1-Benzyl 2-methyl (S)-(−)-1,2-aziridinedicarboxylate, Methyl (S)-N-Cbz-aziridine-2-carboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$472.00

$472.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$472.00

About This Item

Empirical Formula (Hill Notation):
C12H13NO4
CAS Number:
Molecular Weight:
235.24
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$472.00


Check Cart for Availability

Request a Bulk Order

Assay

96%

optical activity

[α]20/D −25°, c = 1 in toluene

refractive index

n20/D 1.519 (lit.)

bp

160-165 °C/2 mmHg (lit.)

density

1.19 g/mL at 25 °C (lit.)

functional group

ester
phenyl

SMILES string

COC(=O)C1CN1C(=O)OCc2ccccc2

InChI

1S/C12H13NO4/c1-16-11(14)10-7-13(10)12(15)17-8-9-5-3-2-4-6-9/h2-6,10H,7-8H2,1H3/t10-,13?/m0/s1

InChI key

GTZJUBQWCWZING-NKUHCKNESA-N

General description

Methyl (S)-(−)-N-Z-aziridine-2-carboxylate is commonly used as an electrophilic aziridination reagent in organic synthesis. It is a chiral bicyclic lactam reagent that can be used for the synthesis of chiral aziridines from alkenes. The reagent is particularly useful in the synthesis of chiral building blocks.

Application

Methyl (S)-(−)-N-Z-aziridine-2-carboxylate can be used as a reactant in the total synthesis of α-C-mannosyltryptophan (C-Man-Trp), a naturally occurring C-glycosylamino acid. It is also used in the preparation of optically active aziridine derivatives.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service