Skip to Content
Merck
All Photos(2)

Key Documents

515736

Sigma-Aldrich

Rhenium(VII) oxide

99.995% trace metals basis

Synonym(s):

Dirhenium heptoxide, Rhenium heptoxide

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$413.00

$413.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$413.00

About This Item

Linear Formula:
Re2O7
CAS Number:
Molecular Weight:
484.41
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

$413.00


Check Cart for Availability

Request a Bulk Order

Assay

99.995% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: rhenium

bp

360 °C (lit.)

mp

220 °C (lit.)

density

6.103 g/mL at 25 °C (lit.)

SMILES string

O=[Re](=O)(=O)O[Re](=O)(=O)=O

InChI

1S/7O.2Re

InChI key

NBGOSNNAAHDRLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Precursor to organorhenium oxides which catalyze olefin oxidation, metathesis and other transformations.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service