All Photos(1)
About This Item
Linear Formula:
CH3OC6H4C6H4Br
CAS Number:
Molecular Weight:
263.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
95%
mp
143-145 °C (lit.)
SMILES string
COc1ccc(cc1)-c2ccc(Br)cc2
InChI
1S/C13H11BrO/c1-15-13-8-4-11(5-9-13)10-2-6-12(14)7-3-10/h2-9H,1H3
InChI key
CMYZTJCWFRFRIW-UHFFFAOYSA-N
Legal Information
Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
A new germanium-based linker for solid phase synthesis of aromatics: Synthesis of a pyrazole library.
Spivey AC, et al.
The Journal of Organic Chemistry, 65(17), 5253-5263 (2000)
Synthesis and ordered aggregation in water of a blue light-emitting PEO-PHP-PEO triblock oligomer.
He H, et al.
Chinese Science Bulletin, 48(15), 1525-1530 (2003)
Fabien Durola et al.
Chemical communications (Cambridge, England), (2)(2), 171-173 (2005-12-24)
Bi-isoquinoline can be used as a building block to prepare a new family of non-sterically hindering chelates, including a macrocyclic system; the endocyclic nature of the ligands has been confirmed by the X-ray structure of an octahedral tris-chelate iron(II) complex
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service