Skip to Content
Merck
All Photos(2)

Key Documents

514292

Sigma-Aldrich

1,1′-Bi-2-naphthol bis(trifluoromethanesulfonate)

97%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$253.00

$253.00


Estimated to ship on16 May 2025


Request a Bulk Order

Select a Size

Change View
25 G
$253.00

About This Item

Linear Formula:
(CF3SO3C10H6-)2
CAS Number:
Molecular Weight:
550.45
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$253.00


Estimated to ship on16 May 2025


Request a Bulk Order

Quality Level

Assay

97%

mp

118-121 °C (lit.)

functional group

fluoro
triflate

SMILES string

FC(F)(F)S(=O)(=O)Oc1ccc2ccccc2c1-c3c(OS(=O)(=O)C(F)(F)F)ccc4ccccc34

InChI

1S/C22H12F6O6S2/c23-21(24,25)35(29,30)33-17-11-9-13-5-1-3-7-15(13)19(17)20-16-8-4-2-6-14(16)10-12-18(20)34-36(31,32)22(26,27)28/h1-12H

InChI key

OYJLCOSEYYZULE-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R D Myers et al.
Psychopharmacologia, 42(2), 109-115 (1975-05-28)
Rats of three different strains were tested systematically for their selection of water or ethanol offered in a two-choice situation. In order that preference-aversion curves could be derived, the concentrations of ethanol were increased in strength from 3 to 30%

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service