Skip to Content
Merck
All Photos(1)

Key Documents

513369

Sigma-Aldrich

Luperox® Delta X-9, 2-Butanone peroxide solution

~31 wt. % in 2,2,4-trimethyl-1,3-pentanediol diisobutyrate

Synonym(s):

2-Butanone peroxide solution, Methyl ethyl ketone peroxide solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C(OOH)(CH3)OOC(OOH)(CH3)C2H5
CAS Number:
Molecular Weight:
210.22
Beilstein:
1759757
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

reaction suitability

reagent type: oxidant

concentration

~31 wt. % in 2,2,4-trimethyl-1,3-pentanediol diisobutyrate

refractive index

n20/D 1.437

density

1.005 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CCC(C)(OO)OOC(C)(CC)OO

InChI

1S/C8H18O6/c1-5-7(3,11-9)13-14-8(4,6-2)12-10/h9-10H,5-6H2,1-4H3

InChI key

WFUGQJXVXHBTEM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Polymerization initiator

Legal Information

Product of Arkema Inc.
Luperox is a registered trademark of Arkema Inc.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Org. Perox. D - Repr. 2 - Skin Corr. 1B

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G V Skúladóttir et al.
Lipids, 29(5), 351-357 (1994-05-01)
Weanling male Sprague-Dawley rats were fed diets for four weeks which differed in their content of n-6 (corn oil; CO) and n-3 fatty acids (fish oil; FO), but were similar in their content of saturated and monounsaturated fatty acids and
Jing Zhang et al.
Journal of hazardous materials, 181(1-3), 1024-1030 (2010-06-18)
Methyl ethyl ketone peroxide (MEKPO) is widely used in polymer industry. It is highly sensitive to heat, friction, shock, flame or other sources of ignition, causing risks in production, storage and transportation. In this article, MEKPO is synthesized at a
M N Bozdemir et al.
Human & experimental toxicology, 30(12), 2002-2006 (2011-04-22)
Although ingestion of methyl ethyl ketone peroxide (MEKP) is rare, it carries a high risk of morbidity and mortality. This paper reports the first such case from Turkey in which a 70-year-old man unintentionally ingested MEKP in his kitchen. The
Jo-Ming Tseng et al.
Journal of hazardous materials, 142(3), 765-770 (2006-08-15)
Methyl ethyl ketone peroxide (MEKPO) is a typical organic peroxide with thermally unstable nature that has been broadly employed in the manufacturing process of acrylic resins, as a hardening agent for fiberglass-reinforced plastics, and as a curing agent for unsaturated
P J Karhunen et al.
Human & experimental toxicology, 9(3), 197-200 (1990-05-01)
Fatal massive peripheral zonal hepatic necrosis developed in a 47-year-old man who accidentally ingested a solution of methyl ethyl ketone peroxide (MEKP) in dimethyl phthalate. Such solutions contain about 10% active oxygen. The clinical course was characterized by temporary cardiac

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service