Skip to Content
Merck
All Photos(1)

Key Documents

512834

Sigma-Aldrich

3-(Hydroxymethyl)phenylboronic acid

≥95%

Synonym(s):

3-Hydroxymethylbenzeneboronic acid

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$354.00

$354.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$354.00

About This Item

Linear Formula:
HOCH2C6H4B(OH)2
CAS Number:
Molecular Weight:
151.96
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$354.00


Check Cart for Availability

Request a Bulk Order

Assay

≥95%

mp

95-99 °C (lit.)

functional group

hydroxyl

SMILES string

OCc1cccc(c1)B(O)O

InChI

1S/C7H9BO3/c9-5-6-2-1-3-7(4-6)8(10)11/h1-4,9-11H,5H2

InChI key

HGTDLKXUWVKLQX-UHFFFAOYSA-N

Application

Reactant involved in:
  • Copper-mediated trifluoromethylation
  • Copper-catalyzed transformations from arylboronic acids in water
  • Mitsunobu, Suzuki, and amidation reactions with hydroxyphenylamino bromopyrazinecarboxylate

Reactant involved in the synthesis of biologically active molecules including:
  • Mycobacterium tuberculosis H37Rv chorismate mutase inhibitors via Suzuki coupling reactions
  • HIV protease inhibitors with antiviral activity against drug-resistant viruses
  • Pyrrole derivatives for use as PDE4B inhibitors

Other Notes

Contains varying amounts of anhydride

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service