Skip to Content
Merck
All Photos(1)

Documents

511943

Sigma-Aldrich

1-Acetyl-4-(4-hydroxyphenyl)piperazine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H16N2O2
CAS Number:
Molecular Weight:
220.27
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

mp

180-185 °C (lit.)

SMILES string

CC(=O)N1CCN(CC1)c2ccc(O)cc2

InChI

1S/C12H16N2O2/c1-10(15)13-6-8-14(9-7-13)11-2-4-12(16)5-3-11/h2-5,16H,6-9H2,1H3

InChI key

AGVNLFCRZULMKK-UHFFFAOYSA-N

General description

1-Acetyl-4-(4-hydroxyphenyl)piperazine (AHPP), a piperazine derivative, is a structural analog of acetaminophen. Its multi-step preparation using bis-(2-chloroethyl)amine hydrochloride and p-aminophenol as starting materials has been reported.2

Application

1-Acetyl-4-(4-hydroxyphenyl)piperazine may be used in the preparation of 1-acetyl-4-(4-octadecyloxyphenyl)piperazine. It may also be used in the multi-step synthesis of ketoconazole, its 1,2,4-triazole and thiazole analogs.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 1, 2,4-triazole and thiazole analogs of ketoconazole.
Huang LF, et al.
Journal of Heterocyclic Chemistry, 34(2), 469-476 (1997)
Design of new potent and selective secretory phospholipase A2 inhibitors. 6-Synthesis, structure-activity relationships and molecular modelling of 1-substituted-4-[4,5-dihydro-1,2,4-(4H)-oxadiazol-5-one-3-yl(methyl)]-functionalized aryl piperazin/one/dione derivatives.
Meddad-Belhabich N, et al.
Bioorganic & Medicinal Chemistry, 18(10), 3588-3600 (2010)
Synthesis of Ketoconazole by Phase-transfer Catalysis [J].
Chen YH and Long SJ.
Chinese Journal of Pharmaceuticals / Chung-kuo i yao kung yeh tsa chih, 8, 005-005 (2008)
New process for synthesis of 1-acetyl-4-(4-hydroxyphenyl) piperazine [J].
Zheng, CZ, et al.
Huaxue Shiji, 10, 27-27 (2010)
S Shi et al.
Biosensors & bioelectronics, 72, 205-210 (2015-05-20)
We propose in this work a general and versatile methodology for electrochemical monitoring of persistent pharmaceutical micropollutants. The system presented is based on an electroactive and electropolymerized hapten (mimetic molecule of the pollutant to be detected) and a specific antibody

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service