Skip to Content
Merck
All Photos(1)

Key Documents

510726

Sigma-Aldrich

2-Cyanobenzophenone

97%

Synonym(s):

2-Benzoylbenzonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$470.00

$470.00


Estimated to ship on26 April 2025


Request a Bulk Order

Select a Size

Change View
1 G
$470.00

About This Item

Linear Formula:
NCC6H4OCC6H5
CAS Number:
Molecular Weight:
207.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$470.00


Estimated to ship on26 April 2025


Request a Bulk Order

Quality Level

Assay

97%

mp

82-87 °C (lit.)

functional group

ketone
nitrile
phenyl

SMILES string

O=C(c1ccccc1)c2ccccc2C#N

InChI

1S/C14H9NO/c15-10-12-8-4-5-9-13(12)14(16)11-6-2-1-3-7-11/h1-9H

InChI key

XPBIJHFBORWDCM-UHFFFAOYSA-N

General description

2-Cyanobenzophenone, also known as 2-benzoylbenzonitrile, is a carbonyl-ene-nitrile compound.[1] Its crystal structure belongs to the monoclinic space group, P21/c.[2] 2-Cyanobenzophenone reacts with furan to form the corresponding adduct.[1] Its reduction under electrochemical condition led to the formation of 2-t-butylbenzophenone, 3-t-butyl-3-phenylphthalide and 6-t-butyl-3-phenylphthalide.[3]

Application

2-Cyanobenzophenone (2-benzoylbenzonitrile) may be used in the preparation of 3-phenyl-3H-isobenzofuran-1-ylidenamine.[4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copper-catalyzed Addition Reactions of Aromatics and Ketones to 2-Aza-2, 4-cyclopentadienone: Facile and Efficient Transformation of Carbonyl-ene-nitriles to 1 H-Pyrrolin-2 (5H)-ones.
Murai M, et al.
The Journal of Organic Chemistry, 73(22), 9174-9176 (2008)
An Efficient Synthesis of 3-Substituted 3H-Isobenzo-furan-1-ylidenamines by the Reaction of 2-Cyano-benzaldehydes with Organolithiums and Their Conversion into Isobenzofuran-1 (3H)-ones.
Kobayashi K, et al.
Heterocycles, 83(1), 99-99 (2011)
Electrochemical t-Butylation of Some Aromatic Ketones.
Kristensen LH and Lund H.
Acta Chemica Scandinavica. Series B, 33, 735-741 (1979)
Structure of 2-cyanobenzophenone.
Preut H, et al.
Acta Crystallographica Section C, Crystal Structure Communications, 48(5), 938-940 (1992)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service