Skip to Content
Merck
All Photos(1)

Key Documents

510262

Sigma-Aldrich

Methallyltrimethylsilane

97%

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
$431.00

$431.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 ML
$431.00

About This Item

Linear Formula:
H2C=C(CH3)CH2Si(CH3)3
CAS Number:
Molecular Weight:
128.29
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$431.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

bp

109-111 °C (lit.)

density

0.73 g/mL at 25 °C (lit.)

SMILES string

CC(=C)C[Si](C)(C)C

InChI

1S/C7H16Si/c1-7(2)6-8(3,4)5/h1,6H2,2-5H3

InChI key

OWVJMAKUWHECNI-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

21.0 °F - closed cup

Flash Point(C)

-6.11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service