497584
BACH-EI™ hydroboration reagent solution
2.0 M in THF
Synonym(s):
Borane N-ethyl-N-isopropylaniline complex
About This Item
reaction suitability
reagent type: reductant
concentration
2.0 M in THF
bp
65-67 °C
density
0.904 g/mL at 25 °C
storage temp.
2-8°C
SMILES string
B.CCN(C(C)C)c1ccccc1
InChI
1S/C11H17N.BH3/c1-4-12(10(2)3)11-8-6-5-7-9-11;/h5-10H,4H2,1-3H3;1H3
InChI key
XYUIGBMTIMBDDQ-UHFFFAOYSA-N
Application
- Hydroboration reactions
- CBS-oxazaborolidine-catalyzed borane reduction
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3
Target Organs
Respiratory system
Supplementary Hazards
Storage Class Code
4.3 - Hazardous materials which set free flammable gases upon contact with water
WGK
WGK 3
Flash Point(F)
-20.0 °F - closed cup
Flash Point(C)
-28.89 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service