Skip to Content
Merck
All Photos(1)

Key Documents

49671

Sigma-Aldrich

(R)-(+)-1,2-Epoxyheptane

purum, ≥97.0% (sum of enantiomers, GC)

Synonym(s):

(R)-2-Pentyloxirane, (R)-Heptene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14O
CAS Number:
Molecular Weight:
114.19
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

grade

purum

Assay

≥97.0% (sum of enantiomers, GC)

refractive index

n20/D 1.415 (lit.)

bp

56 °C/30 mmHg (lit.)

density

0.836 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCCC[C@@H]1CO1

InChI

1S/C7H14O/c1-2-3-4-5-7-6-8-7/h7H,2-6H2,1H3/t7-/m1/s1

InChI key

NMOFYYYCFRVWBK-SSDOTTSWSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

68.0 °F - closed cup

Flash Point(C)

20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service