Skip to Content
Merck
All Photos(2)

Key Documents

495093

Sigma-Aldrich

Magnesium bromide

anhydrous, powder, ≥99.99%

Synonym(s):

Hydrobromic acid magnesium salt, Magnesium dibromide

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$183.00
5 G
$648.00

$183.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$183.00
5 G
$648.00

About This Item

Linear Formula:
MgBr2
CAS Number:
Molecular Weight:
184.11
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
anhydrous
form:
powder

$183.00


Check Cart for Availability

Request a Bulk Order

grade

anhydrous

Quality Level

Assay

≥99.99%

form

powder

impurities

≤100.0 ppm Trace Metal Analysis

mp

711 °C (lit.)

density

3.72 g/mL at 25 °C (lit.)

SMILES string

Br[Mg]Br

InChI

1S/2BrH.Mg/h2*1H;/q;;+2/p-2

InChI key

OTCKOJUMXQWKQG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Other Notes

Metals purity does not include Tungsten which is a by-product of the method of manufacture. Tungsten should not interfere with use of product.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service