Skip to Content
Merck
All Photos(2)

Key Documents

49350

Sigma-Aldrich

Tetrabutylammonium glutaconaldehyde enolate hydrate

≥97.0% (T)

Synonym(s):

2-Pentenedial tetrabutylammonium salt, 5-Hydroxy-2,4-pentadienal tetrabutylammonium salt, Glutaconaldehyde tetrabutylammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3CH2CH2CH2)4N(OCH=CHCH=CHCHO) · xH2O
Molecular Weight:
339.56 (anhydrous basis)
MDL number:
UNSPSC Code:
12352114
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (T)

mp

107-110 °C

SMILES string

O.[O-]\C=C\C=C\C=O.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.C5H6O2.H2O/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;6-4-2-1-3-5-7;/h5-16H2,1-4H3;1-6H;1H2/q+1;;/p-1/b;3-1+,4-2+;

InChI key

PVRWOPYOUJDSKI-PXBPUWEMSA-M

Other Notes

The glutaconaldehyde anion serves as an interesting intermediate for the synthesis of various heterocyclic compounds[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Becher
Synthesis, 589-589 (1980)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service