Skip to Content
Merck
All Photos(1)

Key Documents

492744

Sigma-Aldrich

4-Bromobenzaldehyde dimethyl acetal

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4CH(OCH3)2
CAS Number:
Molecular Weight:
231.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.532 (lit.)

bp

254 °C (lit.)

density

1.383 g/mL at 25 °C (lit.)

functional group

acetal
bromo
ether

SMILES string

COC(OC)c1ccc(Br)cc1

InChI

1S/C9H11BrO2/c1-11-9(12-2)7-3-5-8(10)6-4-7/h3-6,9H,1-2H3

InChI key

TWFNGPDYMFEHOB-UHFFFAOYSA-N

General description

4-Bromobenzaldehyde diethyl acetal is an acetal derivative of 4-bromobenzaldehyde. Its enthalpy of vaporization at boiling point (527.15K) is 46.499kjoule/mol. 4-Bromobenzaldehyde diethyl acetal can be synthesized by reacting 4-bromobenzaldehyde with methanol in the presence of triethylamine and titanium chloride.

Application

4-Bromobenzaldehyde diethyl acetal may be used in the synthesis of [2-(4-dimethoxymethylphenyl)-vinyl]-trimethyl-silane and di(4-formylphenyl)diphenylsilane.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Asymmetric allylation polymerization of bis (allylsilane) and dialdehyde containing arylsilane structure.
Kumagai T and Itsuno S.
Macromolecules, 35(13), 5323-5325 (2002)
Kokorin AI.
Nitroxides - Theory, Experiment and Applications., 362-362 (2012)
Yaws CL.
Thermophysical Properties of Chemicals and Hydrocarbons, 567-567 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service