492000
Sodium butyrate-2,4-13C2
99 atom % 13C
Synonym(s):
Butyric-2,4-13C2 acid sodium salt
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
13CH3CH213CH2CO2Na
CAS Number:
Molecular Weight:
112.07
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
Recommended Products
isotopic purity
99 atom % 13C
form
solid
mp
250-253 °C (lit.)
mass shift
M+2
SMILES string
[Na+].[13CH3]C[13CH2]C([O-])=O
InChI
1S/C4H8O2.Na/c1-2-3-4(5)6;/h2-3H2,1H3,(H,5,6);/q;+1/p-1/i1+1,3+1;
InChI key
MFBOGIVSZKQAPD-CFARNWPNSA-M
Related Categories
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Claudia Reyes-Goya et al.
Toxics, 8(3) (2020-07-24)
Sunitinib (Su) is a tyrosine kinase inhibitor with antiangiogenic and antineoplastic effects that is recommended therapy for renal cell carcinoma, gastrointestinal stromal tumors, and pancreatic neuroendocrine tumors. Arterial hypertension is one of the adverse effects observed in the treatment with
Álvaro Santana-Garrido et al.
Antioxidants (Basel, Switzerland), 9(9) (2020-09-24)
Oxidative stress plays an important role in the pathogenesis of ocular diseases, including hypertensive eye diseases. The beneficial effects of olive oil on cardiovascular diseases might rely on minor constituents. Currently, very little is known about the chemical composition and/or
Tomoya Sasahara et al.
iScience, 24(9), 102936-102936 (2021-08-31)
Amyloid β-protein (Aβ) may contribute to worsening of Alzheimer's disease (AD) through vascular dysfunction, but the molecular mechanism involved is unknown. Using ex vivo blood vessels and primary endothelial cells from human brain microvessels, we show that patient-derived Aβ assemblies, termed
Wen-Hsuan Yang et al.
Molecular cancer research : MCR, 18(1), 79-90 (2019-10-24)
Ovarian cancer is the deadliest gynecologic cancer. Despite recent advances, clinical outcomes remain poor, necessitating novel therapeutic approaches. To investigate metabolic susceptibility, we performed nutrigenetic screens on a panel of clear cell and serous ovarian cancer cells and identified cystine
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service