Skip to Content
Merck
All Photos(1)

Key Documents

49157

Sigma-Aldrich

2-(2-Cyanethyl)cyclohexanone

≥97.0%

Synonym(s):

2-Oxocyclohexane-2-propionitrile, 3-(2-Oxocyclohexyl)propionitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13NO
CAS Number:
Molecular Weight:
151.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0%

refractive index

n20/D 1.474

density

1.024 g/mL at 20 °C (lit.)

SMILES string

O=C1CCCCC1CCC#N

InChI

1S/C9H13NO/c10-7-3-5-8-4-1-2-6-9(8)11/h8H,1-6H2

InChI key

SPTVCXKSSRBTMN-UHFFFAOYSA-N

General description

2-(2-Cyanethyl)cyclohexanone, also known as 3-(2-oxocyclohexyl)propionitrile, is an α-substituted cyclohexanone. It reacts with BrF3(bromine trifluoride) to form the corresponding oxadecaline.

Application

2-(2-Cyanethyl)cyclohexanone may be used in the synthesis of 4-keto-nona-1,9-dioic acid by reacting with hydrogen peroxide.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Purich DL
Enzyme Kinetics and Mechanism, Part F, 354, 109-109 (2002)
Katritzky AR and Talyor RJK
Comprehensive Organic Functional Group Transformations II, 2986-2986 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service