Skip to Content
Merck
All Photos(1)

Key Documents

490482

Sigma-Aldrich

Pentane-d12

98 atom % D

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$1,400.00

$1,400.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$1,400.00

About This Item

Linear Formula:
CD3(CD2)3CD3
CAS Number:
Molecular Weight:
84.22
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

$1,400.00


Check Cart for Availability

Request a Bulk Order

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

form

liquid

technique(s)

NMR: suitable

refractive index

n20/D 1.358 (lit.)

bp

36 °C (lit.)

mp

-130 °C (lit.)

density

0.731 g/mL at 25 °C (lit.)

mass shift

M+12

SMILES string

[2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]

InChI

1S/C5H12/c1-3-5-4-2/h3-5H2,1-2H3/i1D3,2D3,3D2,4D2,5D2

InChI key

OFBQJSOFQDEBGM-HYVJACIRSA-N

General description

Pentane-d12 is a deuterated NMR solvent useful in NMR-based research and analyses.

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-56.2 °F - closed cup

Flash Point(C)

-49.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Flocculation behavior of asphaltenes in solvent/nonsolvent systems.
Ostlund JA, et al.
Journal of Colloid and Interface Science, 253(1), 150-158 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service