Skip to Content
Merck
All Photos(2)

Key Documents

489743

Sigma-Aldrich

Hydroxylamine-15N hydrochloride

≥98 atom % 15N, ≥95% (CP)

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$1,690.00

$1,690.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 G
$1,690.00

About This Item

Linear Formula:
H215NOH · HCl
CAS Number:
Molecular Weight:
70.48
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

$1,690.00


Check Cart for Availability

Request more information

isotopic purity

≥98 atom % 15N

Quality Level

Assay

≥95% (CP)

mp

155-157 °C (lit.)

mass shift

M+1

SMILES string

Cl.[15NH2]O

InChI

1S/ClH.H3NO/c;1-2/h1H;2H,1H2/i;1+1

InChI key

WTDHULULXKLSOZ-IEOVAKBOSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

spleen

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service