Skip to Content
Merck
All Photos(1)

Key Documents

487465

Sigma-Aldrich

1,4-Phenylenediamine-15N2

98 atom % 15N

Synonym(s):

1,4-Benzenediamine-15N2, 1,4-Diamino-15N2-benzene

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$1,690.00

$1,690.00


Check Cart for Availability

Request more information

Select a Size

Change View
250 MG
$1,690.00

About This Item

Linear Formula:
C6H4(15NH2)2
CAS Number:
Molecular Weight:
110.13
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

$1,690.00


Check Cart for Availability

Request more information

isotopic purity

98 atom % 15N

Quality Level

form

solid

bp

267 °C (lit.)

mass shift

M+2

SMILES string

[15NH2]c1ccc([15NH2])cc1

InChI

1S/C6H8N2/c7-5-1-2-6(8)4-3-5/h1-4H,7-8H2/i7+1,8+1

InChI key

CBCKQZAAMUWICA-BFGUONQLSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Sens. 1 - STOT SE 1 Oral

Target Organs

Kidney,Heart,Musculo-skeletal system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service