Skip to Content
Merck
All Photos(1)

Documents

485861

Sigma-Aldrich

L-Alanine-2-d

≥98 atom % D, ≥98% (CP)

Synonym(s):

Deuterated L-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CD(NH2)CO2H
CAS Number:
Molecular Weight:
90.10
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

≥98 atom % D

Assay

≥98% (CP)

form

solid

optical activity

[α]25/D +14.5°, c = 2 in 1 M HCl

mp

314.5 °C (dec.) (lit.)

mass shift

M+1

SMILES string

[2H][C@@](C)(N)C(O)=O

InChI

1S/C3H7NO2/c1-2(4)3(5)6/h2H,4H2,1H3,(H,5,6)/t2-/m0/s1/i2D

InChI key

QNAYBMKLOCPYGJ-MYSWAXPFSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

M Mostafizar Rahman et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 214, 269-276 (2019-02-21)
The non-invasive spectroscopic technique is capable to detect the biomolecular structure spectral features that are associated with biological, nutritional and biodegradation functions. However, to date, no research has been reported on alteration of bioactive compounds/carbohydrate traits on physiochemical and structure
Luciana Louzada Prates et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 188, 495-506 (2017-08-02)
The objectives of this study were to quantify the chemical profile and the magnitude of differences in the oat and barley grain varieties developed by Crop Development Centre (CDC) in terms of Cornell Net Carbohydrate Protein System (CNCPS) carbohydrate sub-fractions:
Robert Moučka et al.
International journal of molecular sciences, 21(22) (2020-11-26)
Polypyrrole one-dimensional nanostructures (nanotubes, nanobelts and nanofibers) were prepared using three various dyes (Methyl Orange, Methylene Blue and Eriochrome Black T). Their high electrical conductivity (from 17.1 to 60.9 S cm-1), good thermal stability (in the range from 25 to
Sweety Sharma et al.
Journal of forensic sciences, 65(3), 751-761 (2019-12-14)
Cosmetic foundation creams are encountered as evidentiary material in criminal investigations, particularly in cases related to sexual and physical assault against women. Analyzing foundation cream exhibit is a challenging task as the exhibit is recovered in trace quantity with similar
Eray Tulukcu et al.
Foods (Basel, Switzerland), 8(4) (2019-04-17)
Six species of Salvia seeds cultivated and grown in Cumra/Konya (Turkey) were evaluated using headspace gas chromatography mass spectroscopy (GC-MS) and Fourier transform infrared spectroscopy-attenuated total reflectance (FTIR-ATR) combined chemometrics of hierarchical cluster analysis (HCA) and principal component analysis (PCA).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service