Skip to Content
Merck
All Photos(1)

Key Documents

485179

Sigma-Aldrich

Acetonitrile-1-13C,15N

99 atom % 13C, 98 atom % 15N

Synonym(s):

Methyl cyanide-13C,15N

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$1,580.00

$1,580.00


Estimated to ship on09 April 2025


Request more information

Select a Size

Change View
250 MG
$1,580.00

About This Item

Linear Formula:
CH313C15N
CAS Number:
Molecular Weight:
43.04
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

$1,580.00


Estimated to ship on09 April 2025


Request more information

isotopic purity

99 atom % 13C
98 atom % 15N

Quality Level

Assay

99% (CP)

refractive index

n20/D 1.344 (lit.)

bp

81-82 °C (lit.)

mp

−48 °C (lit.)

density

0.824 g/mL at 25 °C

mass shift

M+2

SMILES string

C[13C]#[15N]

InChI

1S/C2H3N/c1-2-3/h1H3/i2+1,3+1

InChI key

WEVYAHXRMPXWCK-SUEIGJEOSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service