Skip to Content
Merck
All Photos(1)

Key Documents

485020

Sigma-Aldrich

Gadolinium(III) bromide

anhydrous, powder, 99.99% trace rare earth metals basis

Synonym(s):

Gadolinium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
GdBr3
CAS Number:
Molecular Weight:
396.96
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

anhydrous

Assay

99.99% trace rare earth metals basis

form

powder

reaction suitability

reagent type: catalyst
core: gadolinium

mp

770 °C (lit.)

SMILES string

Br[Gd](Br)Br

InChI

1S/3BrH.Gd/h3*1H;/q;;;+3/p-3

InChI key

KGOKDPWKDBWITQ-UHFFFAOYSA-K

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service