Skip to Content
Merck
All Photos(1)

Key Documents

484717

Sigma-Aldrich

3,3′-Dithiobis(propionitrile)

98%

Synonym(s):

3,3′-Dithiodipropionitrile, 3-[(2-Cyanoethyl)disulfanyl]propanenitrile

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$398.00

$398.00


0 left in stock

Request a Bulk Order

Select a Size

Change View
5 G
$398.00

About This Item

Linear Formula:
S2(CH2CH2CN)2
CAS Number:
Molecular Weight:
172.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$398.00


0 left in stock

Request a Bulk Order

Quality Level

Assay

98%

mp

48-51 °C (lit.)

functional group

disulfide
nitrile

SMILES string

N#CCCSSCCC#N

InChI

1S/C6H8N2S2/c7-3-1-5-9-10-6-2-4-8/h1-2,5-6H2

InChI key

HHEYGEJOMQFDFN-UHFFFAOYSA-N

Application


  • Synthesis and applications in nanocrystal technology: Research detailed the facile synthesis of 5-(2-Mercaptoethyl)-1H-tetrazole, closely related to 3,3′-Dithiobis(propionitrile), and its application in the preparation of water-soluble nanocrystals and gels, demonstrating potential for high-performance polymers and biomedical applications (Voitekhovich et al., 2016).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service