Skip to Content
Merck
All Photos(1)

Key Documents

482161

Sigma-Aldrich

Poly(dimethylsiloxane), hydroxy terminated

viscosity 3,500 cSt 

Synonym(s):

PDMS

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$303.00

$303.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
$303.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$303.00


Check Cart for Availability

Request a Bulk Order

Quality Level

refractive index

n20/D 1.405

viscosity

3,500 cSt

density

0.98 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

C[Si](C)(O)O

InChI key

XCLIHDJZGPCUBT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

We offer PDMS surface modification kit which allows for durable hydrophilization of various polydimethylsiloxane (PDMS) surfaces.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service