Skip to Content
Merck
All Photos(3)

Key Documents

480991

Sigma-Aldrich

Zinc acetylacetonate hydrate

99.995% trace metals basis

Synonym(s):

Bis(2,4-pentanedionato)zinc, Zn(acac)2

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$146.00
25 G
$529.00

$146.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$146.00
25 G
$529.00

About This Item

Linear Formula:
Zn(C5H7O2)2 · xH2O
CAS Number:
Molecular Weight:
263.61 (anhydrous basis)
Beilstein:
4166200
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$146.00


Check Cart for Availability

Request a Bulk Order

Assay

99.995% trace metals basis

form

solid

reaction suitability

reagent type: catalyst

mp

135-138 °C (lit.)

SMILES string

[H]O[H].CC(=O)\C=C(\C)O[Zn]O\C(C)=C/C(C)=O

InChI

1S/2C5H8O2.H2O.Zn/c2*1-4(6)3-5(2)7;;/h2*3,6H,1-2H3;1H2;/q;;;+2/p-2/b2*4-3-;;

InChI key

KUJHAYOLESEVSA-SUKNRPLKSA-L

Looking for similar products? Visit Product Comparison Guide

Application

  • Atmospheric dielectric barrier discharge plasma synthesis of nanoporous zinc oxide films: This study utilized zinc acetylacetonate hydrate as a precursor for synthesizing nanoporous zinc oxide films, which are important in various applications, including sensors and catalysis (QR Liu et al.).
  • High-Performance Solution-Processed Amorphous InGaZnO Thin Film Transistors: Zinc acetylacetonate hydrate was used in the solution process of fabricating thin-film transistors, demonstrating its utility in electronic device fabrication (Y Xie, D Wang, HH Fong, 2018).
  • Developing conductive highly ordered zinc oxide nanorods: The chemical served as a precursor in the hydrothermal growth of zinc oxide nanorods, which are crucial for various nanotechnology applications (SS A Karim et al., 2020).
  • Beta-cyclodextrin-zinc acetylacetonate inclusion complex formation: This research explored the formation of a complex for sustainable nano-carrier applications in corrosion inhibition, highlighting the compound′s versatility in smart material applications (A Dehghani et al., 2020).
  • Nano/microstructure and optical properties of ZnO particles: Focused on the precipitation of zinc oxide particles from zinc acetylacetonate hydrate, which is significant for developing materials with specific optical properties (Ž Petrović et al., 2015).

Features and Benefits

Novel MOCVD precursor for the preparation of high-crystallinity (Zn,Fe)Fe2O4 films and magnetic property measurements of these films.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Few Monolayer Atomic Layer Deposition (ALD) on Surfaces and Interfaces for Energy Applications

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service