Skip to Content
Merck
All Photos(1)

Key Documents

480320

Sigma-Aldrich

Poly[dimethylsiloxane-co-[3-(2-(2-hydroxyethoxy)ethoxy)propyl]methylsiloxane]

viscosity 75 cSt (lit.)

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$242.00

$242.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
$242.00

About This Item

MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$242.00


Check Cart for Availability

Request a Bulk Order

refractive index

n20/D 1.451

Quality Level

viscosity

75 cSt(lit.)

density

1.06 g/mL at 25 °C

SMILES string

C[Si](C)(O)O.OCCOCCOCCC[Si](O)=O

InChI

1S/C7H16O5Si.C2H8O2Si/c8-2-4-12-6-5-11-3-1-7-13(9)10;1-5(2,3)4/h8-9H,1-7H2;3-4H,1-2H3

InChI key

JCKOMIDYRCKNHN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Emulsifier and foaming agent.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Related Content

We offer PDMS surface modification kit which allows for durable hydrophilization of various polydimethylsiloxane (PDMS) surfaces.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service