Skip to Content
Merck
All Photos(1)

Key Documents

479543

Sigma-Aldrich

Platinum(0)-2,4,6,8-tetramethyl-2,4,6,8-tetravinylcyclotetrasiloxane complex solution

0.104 M in methylvinylcyclosiloxanes

Synonym(s):

2,4,6,8-Tetramethyl-2,4,6,8-tetravinyl-1,3,5,7,2,4,6,8-tetroxatetrasilocane platinum

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$209.00

$209.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$209.00

About This Item

Empirical Formula (Hill Notation):
C12H24O4PtSi4
CAS Number:
Molecular Weight:
539.74
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$209.00


Check Cart for Availability

Request a Bulk Order

reaction suitability

core: platinum
reagent type: catalyst

Quality Level

concentration

0.104 M in methylvinylcyclosiloxanes

bp

207 °C

density

1.017 g/mL at 25 °C

SMILES string

[Pt].C[Si]1(O[Si](C)(O[Si](C)(O[Si](C)(O1)C=C)C=C)C=C)C=C

InChI

1S/C12H24O4Si4.Pt/c1-9-17(5)13-18(6,10-2)15-20(8,12-4)16-19(7,11-3)14-17;/h9-12H,1-4H2,5-8H3;

InChI key

PIZSEPSUZMIOQF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Repr. 1B - Resp. Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Do we have the CAS number or information for the methylvinylcyclosiloxanes in the 0.104 M platinum(0)-2,4,6,8-tetramethyl-2,4,6,8-tetravinylcyclotetrasiloxane complex?

    1 answer
    1. The CAS number used for the methylvinylcyclosiloxanes is 68082-23-5, and the product will consist of approximately 95% of 68082-23-5 and about 5% of 68585-32-0.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service