Skip to Content
Merck
All Photos(1)

Documents

479446

Sigma-Aldrich

Bis(ethylcyclopentadienyl)magnesium

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mg(C5H4C2H5)2
CAS Number:
Molecular Weight:
210.60
MDL number:
UNSPSC Code:
12352300

Assay

97%

density

1.912 g/mL at 25 °C (lit.)

SMILES string

[Mg].CC[C]1[CH][CH][CH][CH]1.CC[C]2[CH][CH][CH][CH]2

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Pyr. Liq. 1 - Water-react 2

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service