Skip to Content
Merck
All Photos(2)

Key Documents

477982

Sigma-Aldrich

2,2-Dimethyl-2H-1-benzopyran-6-carbonitrile

97%

Synonym(s):

6-Cyano-2,2-dimethylchromene

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$129.00

$129.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
1 G
$129.00

About This Item

Empirical Formula (Hill Notation):
C12H11NO
CAS Number:
Molecular Weight:
185.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$129.00


Estimated to ship on20 May 2025


Request a Bulk Order

Assay

97%

mp

45-49 °C (lit.)

functional group

nitrile

SMILES string

CC1(C)Oc2ccc(cc2C=C1)C#N

InChI

1S/C12H11NO/c1-12(2)6-5-10-7-9(8-13)3-4-11(10)14-12/h3-7H,1-2H3

InChI key

YDEQIYMIVRCVAH-UHFFFAOYSA-N

General description

2,2-Dimethyl-2H-1-benzopyran-6-carbonitrile (6-Cyano-2,2-dimethyl-2H-1-benzopyran) is a substituted 2,2-dimethyl-2H-1-benzopyran. It can be synthesized by the condensation reaction between 1,1-diethoxy-3-methyl-2-butene and 4-cyanophenol in the presence of pyridine.[1]

Application

2,2-Dimethyl-2H-1-benzopyran-6-carbonitrile (6-Cyano-2,2-dimethylchromene) may be used in the synthesis of 3,4-epoxy-6-cyano-2,2-dimethylchromene via epoxidation.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olefin epoxidation by a (salen) Mn (III) catalyst covalently grafted on glass beads.
Sfrazzetto GT, et al.
Catalysis Science & Technology, 5(2), 673-679 (2015)
Synthesis of 6-cyano-2, 2-dimethyl-2H-1-benzopyran and other substituted 2, 2-dimethyl-2H-1-benzopyrans.
North JT, et al.
The Journal of Organic Chemistry, 60(11), 3397-3400 (1995)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service