Skip to Content
Merck
All Photos(1)

Key Documents

477532

Sigma-Aldrich

5-tert-Butyl-2-hydroxybenzaldehyde

98%

Synonym(s):

5-tert-Butylsalicylaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CC6H3(OH)CHO
CAS Number:
Molecular Weight:
178.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

refractive index

n20/D 1.539 (lit.)

bp

251-252 °C/729 mmHg (lit.)

density

1.039 g/mL at 25 °C (lit.)

functional group

aldehyde

SMILES string

CC(C)(C)c1ccc(O)c(C=O)c1

InChI

1S/C11H14O2/c1-11(2,3)9-4-5-10(13)8(6-9)7-12/h4-7,13H,1-3H3

InChI key

ZVCQQLGWGRTXGC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5-tert-Butyl-2-hydroxybenzaldehyde is a 5-substituted 2-hydroxy aromatic aldehyde. It participates in the synthesis of Schiff base appended porphyrazine.

Application

5-tert-Butyl-2-hydroxybenzaldehyde may be used in the synthesis of the following organic ligands:
  • 5-tert-butyl-2-hydroxybenzaldehyde thiosemicarbazone (THTB)
  • 4-tert-butyl-2-(4,5-dimethyl-1H-imidazol-2-yl)phenol
  • 2-tert-butyl-6-(4,5-diphenyl-1H-imidazol-2-yl)phenol

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of new class composite adsorbent for enhanced palladium (II) detection and recovery.
Awual MR, et al.
Sensors and Actuators B, Chemical, 209, 790-797 (2015)
Porphyrazines as molecular scaffolds: periphery-core spin coupling between metal ions of a schiff base porphyrazine.
Min Zhao et al.
Angewandte Chemie (International ed. in English), 42(4), 462-465 (2003-02-06)
Ligand characteristics and in situ generation of Pd active species towards C-C coupling using series of 2-(1H-imidazol-2-yl) phenols.
Eseola AO, et al.
J. Mol. Catal. A: Chem., 387, 112-122 (2014)
A novel facial composite adsorbent for enhanced copper (II) detection and removal from wastewater.
Awual MR.
Chemical Engineering Journal, 266, 368-375 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service