Skip to Content
Merck
All Photos(1)

Key Documents

476722

Sigma-Aldrich

3-Bromo-1,1,1-trifluoro-2-propanol

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2CH(OH)CF3
CAS Number:
Molecular Weight:
192.96
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
96%

Assay

96%

refractive index

n20/D 1.4 (lit.)

bp

124.5 °C/743 mmHg (lit.)

density

1.861 g/mL at 25 °C (lit.)

SMILES string

OC(CBr)C(F)(F)F

InChI

1S/C3H4BrF3O/c4-1-2(8)3(5,6)7/h2,8H,1H2

InChI key

VBHIIZIQRDVGDH-UHFFFAOYSA-N

General description

3-Bromo-1,1,1-trifluoro-2-propanol (3-bromo-1,1,1-trifluoropropan-2-ol, BTFP) is a brominated saturated alcohol. It behaves as a thiol-reactive trifluoromethyl probe. Its effective chemical shift dispersion under conditions of varying polarity has been studied.[1] The photodissociation dynamics of BTFP has been investigated by various methods.[2][3][4]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

109.4 °F - closed cup

Flash Point(C)

43 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photodissociation dynamics of 3-bromo-1,1,1-trifluoro-2-propanol and 2-(bromomethyl) hexafluoro-2-propanol at 234 nm: Resonance-enhanced multiphoton ionization detection of Br (2Pj).
Indulkar YN, et al.
J. Chem. Phys. , 134(19), 194313-194313 (2011)
A comparison of chemical shift sensitivity of trifluoromethyl tags: optimizing resolution in 19F NMR studies of proteins.
Ye L, et al.
Journal of Biomolecular Nmr, 62(1), 97-103 (2015)
The periodically forced conversion of 2, 3-epoxy-1-propanol to glycerine: A theoretical analysis.
Rehmus P, et al.
J. Chem. Phys. , 78(12), 7241-7251 (1983)
Photodissociation of 3-Bromo-1, 1, 1-trifluoro-2-propanol at 193 nm: Laser-Induced Fluorescence Detection of OH (?''= 0, J'').
Indulkar YN, et al.
The Journal of Physical Chemistry A, 113(30), 8462-8470 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service