Skip to Content
Merck
All Photos(1)

Key Documents

476439

Sigma-Aldrich

2,6-Dimethylphenyl isothiocyanate

98%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$32.40

$32.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$32.40

About This Item

Linear Formula:
(CH3)2C6H3NCS
CAS Number:
Molecular Weight:
163.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$32.40


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

refractive index

n20/D 1.627 (lit.)

bp

247 °C (lit.)

density

1.085 g/mL at 25 °C (lit.)

functional group

isothiocyanate

SMILES string

Cc1cccc(C)c1N=C=S

InChI

1S/C9H9NS/c1-7-4-3-5-8(2)9(7)10-6-11/h3-5H,1-2H3

InChI key

UULUECCNPPJFBU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reactivity of di (azido) bis (phosphine) complexes of Ni (II), Pd (II) and Pt (II) toward organic isothiocyanates: synthesis, structures, and properties of bis (tetrazole-thiolato) and bis (isothiocyanato) complexes.
Kim YJ, et al.
Dalton Transactions, 17, 3357-3364 (2003)
Yaws CL.
Thermophysical Properties of Chemicals and Hydrocarbons, 565-565 (2014)
Preparation of aryl isothiocyanates via protected phenylthiocarbamates and application to the synthesis of caffeic acid (4-isothiocyanato)phenyl ester.
X Zhang et al.
The Journal of organic chemistry, 65(19), 6237-6240 (2000-09-16)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service