Skip to Content
Merck
All Photos(1)

Key Documents

476129

Sigma-Aldrich

Bis(2-ethylhexyl) maleate

90%

Synonym(s):

Bis(2-ethylhexyl) maleate, Di-2-ethylhexyl maleate, Dioctyl maleate

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$97.50

$97.50


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 ML
$97.50

About This Item

Linear Formula:
CH3(CH2)3CH(C2H5)CH2O2CCH=CHCO2CH2CH(C2H5)(CH2)3CH3
CAS Number:
Molecular Weight:
340.50
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$97.50


Available to ship on22 April 2025Details


Request a Bulk Order

Quality Level

Assay

90%

refractive index

n20/D 1.455 (lit.)

density

0.944 g/mL at 25 °C (lit.)

SMILES string

CCCCC(CC)COC(=O)\C=C/C(=O)OCC(CC)CCCC

InChI

1S/C20H36O4/c1-5-9-11-17(7-3)15-23-19(21)13-14-20(22)24-16-18(8-4)12-10-6-2/h13-14,17-18H,5-12,15-16H2,1-4H3/b14-13-

InChI key

ROPXFXOUUANXRR-YPKPFQOOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bis(2-ethylhexyl) maleate is an aliphatic ester which is prepared by catalytic esterification of maleic acid and 2-ethylhexyl alcohol. It is used as a green plasticizer in variety of applications such as adhesives and coatings.[1][2]

Application


  • Structure‐Performance Guided Design of Sustainable Plasticizers from Biorenewable Feedstocks: Explores the use of bis(2-ethylhexyl) maleate in the design of bio-based sustainable plasticizers (C Plass et al., 2021).


Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - STOT RE 2 Oral

Target Organs

Kidney,Liver

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

365.0 °F - closed cup

Flash Point(C)

185 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Some Study on Esterification of Maleic Acid with 2-Ethyl hexyl Alcohol.
Sadek EM.
J. Chem. Soc. Pak., 20, 271-274 (1998)
Hanno C Erythropel et al.
Chemosphere, 134, 106-112 (2015-04-29)
The ubiquitous presence of the plasticizer di (2-ethylhexyl) phthalate (DEHP) in the environment is of concern due to negative biological effects associated with it and its metabolites. In particular, the metabolite mono (2-ethylhexyl) phthalate (MEHP) is a potential endocrine disruptor.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service