Skip to Content
Merck
All Photos(1)

Key Documents

475912

Sigma-Aldrich

Hexyl isothiocyanate

95%

Synonym(s):

1-Isothiocyanatohexane, n-Hexyl mustard oil

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5NCS
CAS Number:
Molecular Weight:
143.25
Beilstein:
1751837
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

refractive index

n20/D 1.493 (lit.)

bp

136-138 °C/96 mmHg (lit.)

density

0.929 g/mL at 25 °C (lit.)

functional group

amine
isothiocyanate

SMILES string

CCCCCCN=C=S

InChI

1S/C7H13NS/c1-2-3-4-5-6-8-7-9/h2-6H2,1H3

InChI key

WXYAXKKXIGHXDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hexyl isothiocyanate (n-hexyl isothiocyanate) is an isothiocyanate derivative. It has been identified as one of the components present in the oil extracted from the Cruciferae family.[1] On heating the mixture of n-hexyl isothiocyanate and m-chlorophenyl isothiocyanate in the temperature range of 190-230°C, n-hexyl isocyanate is obtained.[2]

Application

Hexyl isothiocyanate may be used as a starting reagent in the synthesis of hexyl isocyanate.[3]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Birsa ML, et al.
Science of Synthesis: Houben-Weyl Methods of Molecular Transformations, 134-134 (2014)
Linskens H-F and Jackson JF.
Gas Chromatography/Mass Spectrometry, Volume 3 of Molecular Methods of Plant Analysis, 265-265 (2012)
Toshiro Noshita et al.
European journal of medicinal chemistry, 44(12), 4931-4936 (2009-09-01)
There is increasing demand for novel anti-inflammatory drugs with different mechanisms of action. We synthesized a series of isothiocyanates 2b-h based on 6-(methylsulfinyl)hexyl isothiocyanate (6-MITC) found in the pungent spice Wasabia japonica. Inhibitory activities against in-vitro growth of tumor cells
Moody CJ.
Comprehensive Organic Functional Group Transformations: Synthesis: carbon with two heteroatoms, each attached by a single bond, 5, 1009-1009 (1995)
Krishna Jayant et al.
Cell reports, 26(1), 266-278 (2019-01-04)
Intracellular recordings in vivo remains the best technique to link single-neuron electrical properties to network function. Yet existing methods are limited in accuracy, throughput, and duration, primarily via washout, membrane damage, and movement-induced failure. Here, we introduce flexible quartz nanopipettes (inner

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service