Skip to Content
Merck
All Photos(1)

Documents

475343

Sigma-Aldrich

Thioglycolic acid

≥97%

Synonym(s):

Mercaptoacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2COOH
CAS Number:
Molecular Weight:
92.12
Beilstein:
506166
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.2 (vs air)

Quality Level

vapor pressure

0.4 mmHg ( 25 °C)

Assay

≥97%

autoignition temp.

662 °F

refractive index

n20/D 1.505 (lit.)

bp

96 °C/5 mmHg (lit.)

mp

−16 °C (lit.)

density

1.326 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

OC(=O)CS

InChI

1S/C2H4O2S/c3-2(4)1-5/h5H,1H2,(H,3,4)

InChI key

CWERGRDVMFNCDR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Product of Arkema Inc.

replaced by

Product No.
Description
Pricing

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

266.0 °F - closed cup

Flash Point(C)

130 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Baofu Han et al.
Colloids and surfaces. B, Biointerfaces, 100, 209-214 (2012-07-07)
Fluorescent carbon dots (CDs) were solvothermaly synthesized in water-glycol medium by using glucose as carbon source and then modified with polyethyleneimine (PEI) for the first time to improve fluorescence quality. The as-prepared CDs were monodispersed sphere particles with a diameter
Fabian Hintzen et al.
International journal of pharmaceutics, 436(1-2), 120-126 (2012-06-12)
The aim of this study was to investigate the influence of molar mass of thiolated polymers (thiomers) on their in situ gelling properties. Chitosan-thioglycolic acid (chitosan-TGA) and pectin-cysteine (pectin-Cys) of increasing molar mass were chosen to produce in situ gels
K Gradauer et al.
Journal of controlled release : official journal of the Controlled Release Society, 165(3), 207-215 (2012-12-12)
An ideal oral drug carrier should facilitate drug delivery to the gastrointestinal tract and its absorption into the systemic circulation. To meet these requirements, we developed a thiomer-coated liposomal delivery system composed of 1,2-dipalmitoyl-sn-glycero-3-phosphocholine (DPPC) and a maleimide-functionalized lipid, to
Gizem Ayna et al.
PloS one, 7(6), e40069-e40069 (2012-07-07)
Pathogen-activated and damage-associated molecular patterns activate the inflammasome in macrophages. We report that mouse macrophages release IL-1β while co-incubated with pro-B (Ba/F3) cells dying, as a result of IL-3 withdrawal, by apoptosis with autophagy, but not when they are co-incubated
Heike E Friedl et al.
Biomaterials, 34(32), 7811-7818 (2013-07-28)
It was the purpose of this study to design and evaluate a chitosan derivative as mucoadhesive excipient for vaginal drug delivery systems. The chemical modification of chitosan was achieved by conjugation of thioglycolic acid (TGA) resulting in 1594 μmol thiol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service