Skip to Content
Merck
All Photos(2)

Documents

474991

Sigma-Aldrich

3-Bromo-4-methylthiophene

95%

Synonym(s):

3-Methyl-4-bromothiophene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5BrS
CAS Number:
Molecular Weight:
177.06
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

refractive index

n20/D 1.579 (lit.)

bp

179-183 °C/770 mmHg (lit.)

density

1.584 g/mL at 25 °C (lit.)

SMILES string

Cc1cscc1Br

InChI

1S/C5H5BrS/c1-4-2-7-3-5(4)6/h2-3H,1H3

InChI key

MBUSOPVRLCFJCS-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

172.4 °F - closed cup

Flash Point(C)

78 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Oligothiophenes are important organic electronic materials which can be produced using synthetic intermediates and Suzuki coupling.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service