Skip to Content
Merck
All Photos(1)

Key Documents

474452

Sigma-Aldrich

2,2,3,4,4,4-Hexafluorobutyl acrylate

95%

Synonym(s):

HFBA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCO2CH2CF2CH(F)CF3
CAS Number:
Molecular Weight:
236.11
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

contains

100 ppm 4-tert-butylcatechol as inhibitor

refractive index

n20/D 1.352 (lit.)

bp

40-43 °C/8 mmHg (lit.)

density

1.389 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

FC(C(F)(F)F)C(F)(F)COC(=O)C=C

InChI

1S/C7H6F6O2/c1-2-4(14)15-3-6(9,10)5(8)7(11,12)13/h2,5H,1,3H2

InChI key

LMVLEDTVXAGBJV-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Lars V Bock et al.
Nature communications, 10(1), 4598-4598 (2019-10-12)
mRNA contexts containing a 'slippery' sequence and a downstream secondary structure element stall the progression of the ribosome along the mRNA and induce its movement into the -1 reading frame. In this study we build a thermodynamic model based on
Chitose Maruyama et al.
Biomolecules, 10(5) (2020-05-21)
Many pharmacologically important peptides are bacterial or fungal in origin and contain nonproteinogenic amino acid (NPA) building blocks. Recently, it was reported that, in bacteria, a cyclopropane-containing NPA 1-aminocyclopropanecarboxylic acid (ACC) is produced from the L-methionine moiety of S-adenosyl-L-methionine (SAM)
Mireille Ginésy et al.
Molecules (Basel, Switzerland), 24(4) (2019-03-01)
Increasing demand for L-arginine by the food and pharmaceutical industries has sparked the search for sustainable ways of producing it. Microbial fermentation offers a suitable alternative; however, monitoring of arginine production and carbon source uptake during fermentation, requires simple and
Emmanuel Guntz et al.
Archives of toxicology, 95(3), 925-934 (2021-01-22)
Hyperbaric 2% prilocaine is increasingly used for spinal anesthesia. It is the only local anesthetic metabolized to o-toluidine, a human bladder carcinogen. Increase of o-toluidine hemoglobin adducts, a marker of o-toluidine ability to modify the DNA structure, was described following
Mélanie Viltard et al.
Aging, 11(14), 4783-4800 (2019-07-28)
The naked mole-rat (Heterocephalus glaber) is characterized by a more than tenfold higher life expectancy compared to another rodent species of the same size, namely, the laboratory mouse (Mus musculus). We used mass spectrometric metabolomics to analyze circulating plasma metabolites

Articles

Polymeric materials, with unique refractive index properties, find applications in photonics and optical coatings for various devices.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service