Skip to Content
Merck
All Photos(1)

Key Documents

47309

Sigma-Aldrich

Fmoc-D-Trp(Boc)-OH

≥95.0% (HPLC)

Synonym(s):

Nα-Fmoc-N(in)-Boc-D-tryptophan

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$457.00

$457.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$457.00

About This Item

Empirical Formula (Hill Notation):
C31H30N2O6
CAS Number:
Molecular Weight:
526.58
Beilstein:
7062970
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

$457.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95.0% (HPLC)

form

solid

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

application(s)

peptide synthesis

functional group

Boc
Fmoc

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)n1cc(C[C@@H](NC(=O)OCC2c3ccccc3-c4ccccc24)C(O)=O)c5ccccc15

InChI

1S/C31H30N2O6/c1-31(2,3)39-30(37)33-17-19(20-10-8-9-15-27(20)33)16-26(28(34)35)32-29(36)38-18-25-23-13-6-4-11-21(23)22-12-5-7-14-24(22)25/h4-15,17,25-26H,16,18H2,1-3H3,(H,32,36)(H,34,35)/t26-/m1/s1

InChI key

ADOHASQZJSJZBT-AREMUKBSSA-N

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fayçal Touti et al.
Nature chemical biology, 15(4), 410-418 (2019-03-20)
The use of competitive inhibitors to disrupt protein-protein interactions (PPIs) holds great promise for the treatment of disease. However, the discovery of high-affinity inhibitors can be a challenge. Here we report a platform for improving the affinity of peptide-based PPI
Ngoc-Duc Doan et al.
Journal of peptide science : an official publication of the European Peptide Society, 21(5), 387-391 (2014-11-18)
The solid-phase synthesis of azapeptides possessing a C-terminal aza-residue has been accomplished by a protocol featuring regioselective alkylation of benzhydrylidene-aza-glycinamide and illustrated by the syntheses of [aza-Lys(6)] growth-hormone-releasing peptide-6 analogs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service