Skip to Content
Merck
All Photos(1)

Key Documents

47300

Sigma-Aldrich

o-Fluoro-DL-phenylalanine

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4CH2CH(NH2)COOH
CAS Number:
Molecular Weight:
183.18
Beilstein:
3201186
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95% (NT)
≥98% (HPLC)
≥98%

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

color

white to faint brown

mp

243-246 °C (lit.)

application(s)

cell analysis
peptide synthesis

SMILES string

NC(Cc1ccccc1F)C(O)=O

InChI

1S/C9H10FNO2/c10-7-4-2-1-3-6(7)5-8(11)9(12)13/h1-4,8H,5,11H2,(H,12,13)

InChI key

NYCRCTMDYITATC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in:
  • Studying mechanism of P 450-mediated oxidation and rearrangement
  • Conversion of racemic α-arylalanines to (R)-β-arylalanines
  • Ribosomal translation of unnatural peptides
  • Synthesis of diisopropylpyridine acetamides for use as Kv1.5 potassium channel antagonists
  • Enantioselective hydrolysis of esters for resolution of nonprotein amino acids

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T T Otani et al.
Journal of pharmaceutical sciences, 71(2), 214-216 (1982-02-01)
Twelve derivatives of 0-fluoro-dl-phenylalanine containing fluorine, chlorine, methoxy, and nitro radicals in various positions of the aromatic ring of the benzoyl group were prepared and tested in a Lactobacillus casei system. It was found that most substitutions in the benzoyl
Elfriede Pittler et al.
Electrophoresis, 30(16), 2897-2904 (2009-08-06)
This work deals with the application of silica-based ligand-exchange chiral stationary phases (CSPs) for the enantioseparation of underivatized amino acids, alpha-hydroxy acids, and dipeptides with packed CEC. Two different possibilities of preparing silica-based CSPs are presented. One phase contains L-4-hydroxyproline
H Nakamichi et al.
Nuclear medicine and biology, 21(7), 959-962 (1994-10-01)
The anabolism of isotopically labeled amino acids was compared between the cerebrum and the cerebellum in conscious rat at three feeding conditions. After L-[2-18F]fluorophenylalanine and L-[2,6-3H]phenylalanine injections, the incorporation rate of both radioactivity into protein fraction showed no difference between
Johnny Castillo Meleán et al.
Organic & biomolecular chemistry, 9(3), 765-769 (2010-11-23)
2-[(18)F]Fluoro-L-phenylalanine and 2-[(18)F]fluoro-L-tyrosine have been developed as promising radiopharmaceuticals for molecular imaging using positron emission tomography (PET). However, the lack of a convenient radiosynthetic pathway has limited their practical use. In this work a new three-step nucleophilic synthesis of these
Kensuke Okuda et al.
Biochemistry, 44(17), 6675-6684 (2005-04-27)
The ribosome-catalyzed peptidyl transferase reaction displays a complex pH profile resulting from two functional groups whose deprotonation is important for the reaction, one within the A-site substrate and a second unidentified group thought to reside in the rRNA peptidyl transferase

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service