Skip to Content
Merck
All Photos(1)

Documents

47296

Sigma-Aldrich

o-Fluoro-L-phenylalanine

puriss., ≥99.0% (sum of enantiomers, HPLC)

Synonym(s):

2-Fluoro-L-phenylalanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10FNO2
CAS Number:
Molecular Weight:
183.18
Beilstein:
2805791
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

puriss.

Assay

≥99.0% (sum of enantiomers, HPLC)

optical activity

[α]20/D −14±1°, c = 1% in H2O

optical purity

enantiomeric ratio: ≥99.5:0.5 (TLC)

SMILES string

N[C@@H](Cc1ccccc1F)C(O)=O

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Kubota et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 37(2), 320-325 (1996-02-01)
L-[methyl-11C]methionine (11C-Met) is a useful tracer for tumor imaging with PET. The drawbacks include a short half-life and high physiological accumulation in abdominal organs. To overcome these shortfalls, the feasible use of [18F]fluorophenylalanine (18F-Phe), which shares the same amino acid
Johnny Castillo Meleán et al.
Organic & biomolecular chemistry, 9(3), 765-769 (2010-11-23)
2-[(18)F]Fluoro-L-phenylalanine and 2-[(18)F]fluoro-L-tyrosine have been developed as promising radiopharmaceuticals for molecular imaging using positron emission tomography (PET). However, the lack of a convenient radiosynthetic pathway has limited their practical use. In this work a new three-step nucleophilic synthesis of these
Kensuke Okuda et al.
Biochemistry, 44(17), 6675-6684 (2005-04-27)
The ribosome-catalyzed peptidyl transferase reaction displays a complex pH profile resulting from two functional groups whose deprotonation is important for the reaction, one within the A-site substrate and a second unidentified group thought to reside in the rRNA peptidyl transferase
H Ito et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 36(7), 1232-1237 (1995-07-01)
Neutral amino acids (NAAs) are transported from the blood to the brain using the same carrier system in a competitive fashion. The purpose of this study is to establish a method for evaluating neutral amino acid transport at the blood-brain
J Hatazawa et al.
Annals of nuclear medicine, 8(3), 213-217 (1994-08-01)
We studied the brain uptake of amino acid in a patient with acute cerebral infarction with L-[2-(F-18)]fluorophenylalanine and positron emission tomography. The increased accumulation of the ligand was specifically found in the peri-infarct area where oxygen metabolism was still maintained

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service