Skip to Content
Merck
All Photos(1)

Key Documents

472565

Sigma-Aldrich

Diethyl cyanophosphonate

90%

Synonym(s):

DEPC, Diethyl phosphoryl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2O)2P(O)CN
CAS Number:
Molecular Weight:
163.11
Beilstein:
1768938
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

90%

refractive index

n20/D 1.403 (lit.)

bp

104-105 °C/19 mmHg (lit.)

density

1.075 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOP(=O)(OCC)C#N

InChI

1S/C5H10NO3P/c1-3-8-10(7,5-6)9-4-2/h3-4H2,1-2H3

InChI key

ZWWWLCMDTZFSOO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethyl cyanophosphonate (DCNP, a Tabun mimic,[1] DECP[2]) is a nerve agent simulant. Its detection by using fluorescein, a reversible fluorescent sensor, has been reported.[2] Its degradation in the presence of suitable organocatalysts (different amines, aminoalcohols and glycols) has been studied.[1]

Application

Coupling reagent for peptide synthesis.[3] Reagent for the phosphorylation of phenols.[4] Activating agent for fluorescent derivatization of carboxylic acids which allows separation by HPLC.[5]

Other Notes

Contains triethyl phosphate

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

176.0 °F - closed cup

Flash Point(C)

80 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T C Bruice et al.
Proceedings of the National Academy of Sciences of the United States of America, 89(5), 1700-1704 (1992-03-01)
The structures of the compounds we call 3a, 3b, and 3c-compounds that incorporate (i) the tripyrrole peptide of the minor-groove-binding distamycin class of compounds and (ii) polyamine ligands that extend from the minor groove and can interact with phosphodiester bonds--were
Jingfeng Rong et al.
Cell cycle (Georgetown, Tex.), 19(12), 1478-1491 (2020-05-07)
As some evidence has demonstrated the role of microRNA-221 (miR-221) on coronary heart disease (CHD), the aim of the present study was to investigate the effect of miR-221-3p on CHD via regulating NLRP3/ASC/pro-caspase-1 inflammasome pathway. Sixty CHD patients and 60
Farkhondeh Fathi et al.
Langmuir : the ACS journal of surfaces and colloids, 27(19), 12098-12105 (2011-08-27)
A simple electrochemical method has been proposed for the preparation of silver nanoparticles that can be used for the detection of cyanide. Both the electrochemical behavior and morphology of the Ag nanoparticles have been characterized in the presence of KCN
Synthetic Communications, 27, 3035-3035 (1997)
Yoon Jeong Jang et al.
The Analyst, 139(7), 1614-1617 (2014-02-22)
Herein, we present fluorescein as a reversible fluorescent sensor for nerve agent simulants diethylchlorophosphate (DCP), diethyl methylphosphonate (DEMP), and diethyl cyanophosphonate (DECP). The superoxide allows for an "off-on" mechanism to regenerate fluorescein. The order of decrease in fluorescence intensity for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service