Skip to Content
Merck
All Photos(1)

Key Documents

472239

Sigma-Aldrich

2,3,4-Trifluorophenyl isocyanate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F3C6H2NCO
CAS Number:
Molecular Weight:
173.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.478 (lit.)

bp

43 °C/4.5 mmHg (lit.)

density

1.432 g/mL at 25 °C (lit.)

SMILES string

Fc1ccc(N=C=O)c(F)c1F

InChI

1S/C7H2F3NO/c8-4-1-2-5(11-3-12)7(10)6(4)9/h1-2H

InChI key

LSKOUPSEWMJCEP-UHFFFAOYSA-N

General description

2,3,4-Trifluorophenyl isocyanate is an aryl fluorinated building block.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaws CL.
The Yaws Handbook of Physical Properties for Hydrocarbons and Chemicals null
Hermecz I, et al.
Advances in Heterocyclic Chemistry, 54 (1992)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service