Skip to Content
Merck
All Photos(2)

Key Documents

471429

Sigma-Aldrich

(S)-(−)-α-Amino-γ-butyrolactone hydrobromide

99%

Synonym(s):

L-Homoserine lactone hydrobromide

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$242.00

$242.00


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
5 G
$242.00

About This Item

Empirical Formula (Hill Notation):
C4H7NO2 · HBr
CAS Number:
Molecular Weight:
182.02
Beilstein:
3912375
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$242.00


Available to ship on22 April 2025Details


Request a Bulk Order

Assay

99%

form

solid

optical activity

[α]20/D −21°, c = 1 in H2O

mp

225 °C (dec.) (lit.)

functional group

ester

storage temp.

2-8°C

SMILES string

Br[H].N[C@H]1CCOC1=O

InChI

1S/C4H7NO2.BrH/c5-3-1-2-7-4(3)6;/h3H,1-2,5H2;1H/t3-;/m0./s1

InChI key

MKLNTBLOABOJFZ-DFWYDOINSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(S)-(-)-α-Amino-γ-butyrolactone hydrobromide may be used as a building block to prepare:
  • β-ketoamide N-acylated-L-homoserine lactones (AHLs) as quorum sensing molecules[1]
  • p-coumaroyl-HSL (homoserine lactone)[2]
  • calpain and lipid peroxidation inhibitors[3]
Used to prepare pseudopeptide inhibitors for Ras farnesyl-protein transferase[4] as well as selenomethionine.[5]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel dual inhibitors of calpain and lipid peroxidation.
Auvin S, et al.
Bioorganic & Medicinal Chemistry Letters, 14(14), 3825-3828 (2004)
Robust routes for the synthesis of N-acylated-L-homoserine lactone (AHL) quorum sensing molecules with high levels of enantiomeric purity.
Hodgkinson JT, et al.
Tetrahedron Letters, 52(26), 3291-3294 (2011)
A new class of homoserine lactone quorum-sensing signals.
Schaefer AL, et al.
Nature, 454(7204), 595-599 (2008)
Koch, T. Buchardt, O.
Synthesis, 1065-1065 (1993)
S L Graham et al.
Journal of medicinal chemistry, 37(6), 725-732 (1994-03-18)
Inhibitors of Ras farnesyl-protein transferase are described. These are reduced pseudopeptides related to the C-terminal tetrapeptide of the Ras protein that signals farnesylation. Deletion of the carbonyl groups between the first two residues of the tetrapeptides either preserves or improves

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service