Skip to Content
Merck
All Photos(1)

Key Documents

471380

Sigma-Aldrich

(R)-(−)-2-(2,5-Dihydrophenyl)glycine

98%

Synonym(s):

(R)-α-Amino-1,4-cyclohexadiene-1-acetic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$65.50

$65.50


Estimated to ship on16 April 2025


Request a Bulk Order

Select a Size

Change View
5 G
$65.50

About This Item

Empirical Formula (Hill Notation):
C8H11NO2
CAS Number:
Molecular Weight:
153.18
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

$65.50


Estimated to ship on16 April 2025


Request a Bulk Order

Quality Level

Assay

98%

form

solid

optical activity

[α]20/D −154°, c = 1 in H2O

reaction suitability

reaction type: solution phase peptide synthesis

mp

280 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

N[C@@H](C(O)=O)C1=CCC=CC1

InChI

1S/C8H11NO2/c9-7(8(10)11)6-4-2-1-3-5-6/h1-2,5,7H,3-4,9H2,(H,10,11)/t7-/m1/s1

InChI key

JBJJTCGQCRGNOL-SSDOTTSWSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service