Skip to Content
Merck
All Photos(1)

Key Documents

470961

Sigma-Aldrich

2,2,3,3,3-Pentafluoropropyl acrylate

98%, contains 100 ppm 4-tert-butylcatechol as inhibitor

Synonym(s):

PFPA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCO2CH2CF2CF3
CAS Number:
Molecular Weight:
204.09
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

contains

100 ppm 4-tert-butylcatechol as inhibitor

refractive index

n20/D 1.336 (lit.)

bp

50 °C/100 mmHg (lit.)

density

1.32 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

FC(F)(F)C(F)(F)COC(=O)C=C

InChI

1S/C6H5F5O2/c1-2-4(12)13-3-5(7,8)6(9,10)11/h2H,1,3H2

InChI key

JDVGNKIUXZQTFD-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

73.4 °F - closed cup

Flash Point(C)

23 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Polymeric materials, with unique refractive index properties, find applications in photonics and optical coatings for various devices.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service