All Photos(1)
About This Item
Linear Formula:
O2NC6H4SO2C6H5
CAS Number:
Molecular Weight:
263.27
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Assay
97%
mp
145-148 °C (lit.)
SMILES string
[O-][N+](=O)c1ccccc1S(=O)(=O)c2ccccc2
InChI
1S/C12H9NO4S/c14-13(15)11-8-4-5-9-12(11)18(16,17)10-6-2-1-3-7-10/h1-9H
InChI key
GKNMUCPEXSCGKC-UHFFFAOYSA-N
General description
2-Nitrophenyl phenyl sulfone (NPPS) is a substituted diarylsulfone derivative. It has been reported to protect the human CEM-SS lymphoblastoid cells from the cytopathic effects of HIV-1 in cell culture. Structure-activity relationship of 2-nitrophenyl phenyl sulfone has been studied.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
J B McMahon et al.
Antimicrobial agents and chemotherapy, 37(4), 754-760 (1993-04-01)
A series of variously substituted diarylsulfones and related derivatives were found to prevent human immunodeficiency virus type 1 (HIV-1) replication and HIV-1-induced cell killing in vitro. One of the more potent derivatives, 2-nitrophenyl phenyl sulfone (NPPS), completely protected human CEM-SS
M Artico et al.
Archiv der Pharmazie, 328(3), 223-229 (1995-03-01)
Various aryl 1-pyrryl sulfones were synthesized and tested as inhibitors of HIV-1. 2-Nitrophenyl-2-ethoxycarbonyl-1-pyrryl sulfone, the most active among test derivatives, was selected as lead compound of the aryl pyrryl sulfone series. The in vitro anti-HIV-1 activity and cytotoxicity of 41
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service