468568
Hexyllithium solution
2.3 M in hexane
Synonym(s):
1-Hexyllithium, n-Hexyllithium, Hexane - lithium complex
About This Item
Recommended Products
form
liquid
Quality Level
concentration
2.3 M in hexane
bp
69 °C
density
0.708 g/mL at 25 °C
storage temp.
2-8°C
SMILES string
[Li]CCCCCC
InChI
1S/C6H13.Li/c1-3-5-6-4-2;/h1,3-6H2,2H3;
InChI key
RMKNFFPDYCVCDI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
- Ni-catalyzed cross-coupling with aryl halides.
- Cu-catalyzed enantioselective allylic alkylation.
Packaging
Legal Information
Signal Word
Danger
Hazard Statements
Hazard Classifications
Aquatic Chronic 2 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Repr. 2 - Skin Corr. 1A - STOT RE 2 Inhalation - STOT SE 3 - Water-react 1
Target Organs
Central nervous system, Nervous system
Supplementary Hazards
Storage Class Code
4.2 - Pyrophoric and self-heating hazardous materials
WGK
WGK 3
Flash Point(F)
15.8 °F - closed cup
Flash Point(C)
-9 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service